Please login to your online account to display your discounted pricing
American Educational Products Premium Geology Collections
Superb specimens carefully selected to illustrate ideas about their formation
$137.70 - $264.60
Specifications
Includes Teacher's guide; reproducible student activity “field notebook” and reference cards
Quantity 12 Specimens
Specimen Size 5.1 x 7.6cm
View More Specs
Includes:  Teacher's guide, reproducible student activity “field notebook,” and reference cards
Products
Catalog Number Mfr. No. Grade Specimen Type Price Quantity    
S67117A
american educational products
2810*
6 to 12 Rocks Each for $137.70
S67116A
american educational products
2870**
5 to 12 Sedimentary Each for $264.60
S67114A
american educational products
2860*
5 to 12 Igneous Each for $264.60
S67115A
american educational products
2880
5 to 9 Metamorphic Each for $264.60
Description & Specifications
Specifications
Includes Teacher's guide; reproducible student activity “field notebook” and reference cards
Quantity 12 Specimens
Specimen Size 5.1 x 7.6cm
  • High quality box contains 12 large specimens in display tray
  • Each measures approximately 2 x 3 in. (5.1 x 7.6cm)
  • Four quality hand lenses and a color identification chart provide clues and answers