Please login to your online account to display your discounted pricing

Invitrogen™ Ambion™ ULTRAhyb™ Ultrasensitive Hybridization Buffer with Manual DFS Item


Proprietary blend of hybridization accelerators and blocking agents to enhance the levels of hybridization

Manufacturer: invitrogen™  LSAM8670M

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. AM8670M

  • / Each
 

Description & Specifications

Specifications

Buffer Hybridization
Storage Requirements 4°C
Quantity 125mL

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.