missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Ambion™ ULTRAhyb™ Ultrasensitive Hybridization Buffer with Manual DFS Item

Proprietary blend of hybridization accelerators and blocking agents to enhance the levels of hybridization

$198.00 - $604.00

Specifications

Buffer Hybridization
Storage Requirements 4°C
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AM8670M Invitrogen™
LSAM8670M
125mL Each for $198.00
AM8669M Invitrogen™
LSAM8669M
4 × 125mL Each for $604.00
Specifications

Description

  • Signals that once took days to visualize are now apparent in hours
  • It is supplied in four bottles containing 125mL each
  • Increase the sensitivity of any blot hybridization experiment
  • Increase random-primed DNA probe sensitivity up to 100x; RNA probe sensitivity 20x
  • Decrease hybridization time to 2 hours
  • Using standard hybridization buffers, only 1 to ;5% of target molecules on a blot hybridizes to probe, making blots a relatively insensitive method for nucleic acid analysis (Vernier et al. (1996) Anal Biochem 235: 11-19): With ULTRAhyb™ Ultrasensitive Hybridization Buffer, the hybridization reaction approaches completion, so that as few as 10,000 molecules can be detected
  • Compatible with RNA and DNA probes labeled isotopically and nonisotopically in northerns, Southerns, and dot/slot blots
  • Contains 50% formamide and is compatible with positively charged membranes
  • Requires 10-100x less nonisotopic DNA probe than standard hybridization buffers, making it possible to perform many more hybridizations with a single labeling reaction

Applications:

DNA and RNA Purification and Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis and Blotting

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

Hybridization
4°C