Please login to your online account to display your discounted pricing

Ambion™ TRIzol™ LS Reagent DFS Item


Recovery RNA, DNA, and protein from a single liquid sample

Manufacturer: life technologies  LS10296028

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10296028

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Cloning, Northern Blotting, Nuclease Protection Assays, RT-PCR (Endpoint), Real-Time PCR, cDNA Library Construction
No. of Samples 200.0
Isolation Technology Organic Extraction
Volume (Metric) 200mL
Quantity 200mL
Sample Size 250μL to 1mL liquid samples (suggested range; scalable)
Sample Type Blood, Liquid Samples (e.g. Serum), Viral Samples
Target Total RNA, Transcriptome RNA, miRNA
Yield 8mg
Final Product Type Total RNA, Transcriptome RNA, micro RNA
Test Time 1 hr
Quantity 200mL