Please login to your online account to display your discounted pricing

Invitrogen™ Ambion™ Tris (1M), pH 8.0 DFS Item


Ready-to-use, certified RNase-free

Manufacturer: invitrogen™  LSAM9855G

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. AM9855G

  • / Each
 

Description & Specifications

Specifications

Quantity 100mL
Storage Requirements Store at room temperature.
Concentration 1M
Concentration 1M