Please login to your online account to display your discounted pricing

Invitrogen™ Ambion™ THE RNA Storage Solution DFS Item


Features to minutesimize base hydrolysis of RNA

Manufacturer: invitrogen™  LSAM7001

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. AM7001

  • / Each
 

Description & Specifications

Specifications

Concentration 1M
Storage Requirements Store at -20°C. Shipped at ambient temperature.
Quantity 50mL
Concentration 1M