Please login to your online account to display your discounted pricing

Ambion™ T7 RNA Polymerase (cloned) 20U/μL DFS Item


A cloned high-purity RNA polymerase with a high specificity for its promoter

Manufacturer: life technologies  LSAM2084

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. AM2084

  • / Each
 

Description & Specifications

Specifications

Sample Type DNA
Includes One tube containing 30,000 units at a concentration of 20U/μL
Quantity 30,000 units
Concentration 20U/μL
For Use With (Application) Synthesizing high specific activity RNA hybridization probes, biologically active mRNA, and antisense RNA
Storage Requirements Store at -20deg.C