Please login to your online account to display your discounted pricing

Invitrogen™ Ambion™ T7 RNA Polymerase (cloned) 20U/μL DFS Item

A cloned high-purity RNA polymerase with a high specificity for its promoter

$627.90 - $627.90

Specifications

For Use With (Application) Synthesizing high specific activity RNA hybridization probes, biologically active mRNA, and antisense RNA
Sample Type DNA
Storage Requirements Store at -20deg.C
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Includes Quantity Price Quantity    

AM2084

DFS Item
 
life technologies
LSAM2084
One tube containing 30,000 units at a concentration of 20U/μL 30,000 units Each for $627.90
Description & Specifications

Specifications

For Use With (Application) Synthesizing high specific activity RNA hybridization probes, biologically active mRNA, and antisense RNA
Sample Type DNA
Storage Requirements Store at -20deg.C

  • This enzyme will transcribe large amounts of RNA from DNA sequences downstream of the promoter and show no cross talk between promoters
  • One tube containing 5000U at a concentration of 20U/μL is provided. Its high purity makes it ideal for synthesizing high specific activity RNA hybridization probes, biologically active mRNA, and antisense RNA
  • The RNA Polymerase is rigorously tested for contaminating nonspecific endonuclease, exonuclease, protease, and RNase activity
  • It is also tested for functionality with the MAXIscript Kit at 20U per reaction
  • Unit Definition: One unit of RNA Polymerase will catalyze the incorporation of 1nmol of nucleoside triphosphate into acid-insoluble material in 60 minutes at 37°C

DNA & RNA Purification & Analysis, Gene Expression Analysis & Genotyping, in vitro Transcription, Nucleic Acid Labeling & Oligo Synthesis, RNA Labeling

Shipping Condition: Dry ice