missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Ambion™ T4 RNA Ligase (cloned) 5U/μL DFS Item

Catalyzes formation of a phosphodiester linkage between a 5'-phosphoryl-terminated nucleic acid donor and a 3'-hydroxyl-terminated nucleic acid acceptor

$327.00 - $327.00

Specifications

For Use With (Application) Mapping studies (RACE), 3'-end labeling RNA molecules, and circularizing RNA and DNA molecules
Sample Type RNA, DNA, and oligonucleotides
Storage Requirements Store at -20deg.C
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Quantity Price Quantity  
Catalog Number Mfr. No. Includes Quantity Price Quantity  
AM2141 Life Technologies
LSAM2141
T4 RNA Ligase and 10X Reaction Buffer 2500 units Each for $327.00
Specifications

Description

  • Substrates include RNA, DNA, and oligonucleotides. One tube containing 500U (5U/μL) is provided along with 10X Reaction Buffer
  • The enzyme can be used for tagging the 5' ends of mRNA with oligonucleotides for mapping studies (RACE), 3'-end labeling RNA molecules, and circularizing RNA and DNA molecules
  • Unit Definition: One unit catalyzes the formation of 1nmol of [5'-32P]-rA12-18 into a phosphatase-resistant form in 30 minutes at 37°C

DNA & RNA Purification & Analysis, Nucleic Acid Labeling & Oligo Synthesis, Oligo Labeling, RNA Labeling

Order Info

Shipping Condition: Dry ice

Description & Specifications

Specifications

Mapping studies (RACE), 3'-end labeling RNA molecules, and circularizing RNA and DNA molecules
RNA, DNA, and oligonucleotides
Store at -20deg.C