Please login to your online account to display your discounted pricing
Invitrogen™ Ambion™ RPA III™ Ribonuclease Protection Assay Kit DFS Item
Fast single-tube reaction
Manufacturer: invitrogen™  LSAM1414
 View more versions of this product
 Disclaimers
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. AM1414
  • / Each
Description & Specifications
Specifications
Includes Kit without manual
Quantity 100 reactions
Storage Requirements Store Hybridization III Buffer, RNase Digestion III Buffer, Yeast RNA, RNase Inactivation/Precipitation III Solution, Gel Loading Buffer II, pTRI-Actin-Mouse, RNase A/RNase T1 Mix, RNase T1, and 5 M Ammonium Acetate should be stored at –20°C. Mouse Liver RNA should be stored at below –70°C. Probe Elution Buffer should be stored at 4°C.