Please login to your online account to display your discounted pricing
Invitrogen™ Ambion™ RPA III™ Ribonuclease Protection Assay Kit DFS Item
Fast single-tube reaction
Manufacturer: invitrogen™  LSAM1415M
 View more versions of this product
 Disclaimers
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. AM1415M
  • / Each
Description & Specifications
Specifications
Includes Kit with manual
Quantity 35 reactions
Storage Requirements Store Hybridization III Buffer, RNase Digestion III Buffer, Yeast RNA, RNase Inactivation/Precipitation III Solution, Gel Loading Buffer II, pTRI-Actin-Mouse, RNase A/RNase T1 Mix, RNase T1, and 5 M Ammonium Acetate at -20°C. Store Mouse Liver RNA at below -70°C. Store Probe Elution Buffer at 4°C.