Please login to your online account to display your discounted pricing
Ambion™ RNaseAlert™ QC System DFS Item
Detects RNase activity in convenient and sensitive fluorimetric assay with real time results
Manufacturer: life technologies  LSAM1966M
 View more versions of this product
 Disclaimers
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. AM1966M
  • / Each
Description & Specifications
Specifications
Product Type QC System with manual
Reagents RNase Control Reagent
Quantity 480 assays