Invitrogen™ Ambion™ RNaseAlert™ QC System DFS Item
Detects RNase activity in convenient and sensitive fluorimetric assay with real time results
$886.20 - $976.00
Specifications
Quantity 480 assays
Reagents RNase Control Reagent
View More Specs
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Products ${productFamilyLength}
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
AM1966M Life Technologies
LSAM1966M
QC System with manual Each for $886.20
AM1966 Life Technologies
LSAM1966
QC System Each for $976.00
Specifications
Description
Ambion™ RNaseAlert™ QC System is a patent-pending technology detects RNase activity in a convenient and sensitive fluorimetric assay that delivers results in real time.
  • Contain sufficient reagents for 5 × 96 high throughput assays
  • Simple, straightforward 30-minute assay
  • Monitor RNase activity in column fractions during protein purification
  • Use with DNaseAlert™ QC System to quantitatively detect both RNases and DNases in the same sample
  • Lypholized
  • RNaseAlert is also fully compatible with Ambion DNaseAlert™ QC System (Cat. No. AM1970)
  • Since DNaseAlert Substrate contains fluorescent tag that is spectrally distinct from RNaseAlert, the two kits can be used simultaneously with a fluorometer for quantitative real-time analyses of both RNases and DNases within single sample
DNA and RNA Purification and Analysis, General RNA Purification Reagents and Accessories, PCR and Real-Time PCR, RNA Extraction, Real Time PCR (qPCR), Total RNA Isolation, Total RNA from Animal Cells and Tissues, Transcriptome RNA Isolation, mRNA Isolation, microRNA Isolation
Order Info
Shipping Condition: Dry ice
Description & Specifications
Specifications
480 assays
RNase Control Reagent