Please login to your online account to display your discounted pricing
Ambion™ RNaseAlert™ Lab Test Kit DFS Item
Ensures that solutions, tubes, tips, etc. are RNase-free
Manufacturer: life technologies  AM1964M
 View more versions of this product
Catalog No. AM1964M
  • / Each
Description & Specifications
Specifications
Product Type Test Kit with manual
Reagents RNase Control Reagent
Quantity 25 reactions
Ambion™ RNaseAlert™ Lab Test kit is a Patent-pending technology detects RNase activity in convenient and sensitive assay that delivers results in real time. Suitable for testing small sample numbers and can be used to ensure that solutions, tubes, tips, etc. are RNase-free; kit contains sufficient reagents for 25 reactions.
  • Detects as little as 3.5 x 10-7 units (∽0.5pg) of RNase A with a fluorometer or by eye
  • Simple, straightforward 30-minute assay
  • Monitor RNase activity in column fractions during protein purification
  • Use with DNaseAlert™ to quantitatively detect both RNases and DNases in the same sample
  • Lypholized
  • RNaseAlert QC System is also available for high-throughput assays in 96-well format (Cat. No. AM1966)
  • RNaseAlert is compatible with Ambion DNaseAlert QC System (Cat. No. AM1970)
  • DNaseAlert Substrate contains fluorescent tag that spectrally distinct from RNaseAlert, two kits can be used simultaneously with a fluorometer for quantitative real-time analyses of both RNases and DNases within single sample
DNA and RNA Purification and Analysis, General RNA Purification Reagents and Accessories, PCR and Real-Time PCR, RNA Extraction, Real Time PCR (qPCR), Total RNA Isolation, Transcriptome RNA Isolation, mRNA Isolation, microRNA Isolation