Invitrogen™ RNaseAlert™ Lab Test Kit DFS Item
Ensures that solutions, tubes, tips, etc. are RNase-free
$271.00 - $271.00
Specifications
Quantity 25 reactions
Reagents RNase Control Reagent
View More Specs
Products 1
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
AM1964 View Documents Invitrogen™
LSAM1964
Test Kit Each for $271.00
Description
Description
Invitrogen™ RNaseAlert™ Lab Test kit is a Patent-pending technology detects RNase activity in a convenient and sensitive assay that delivers results in real time. Suitable for testing small sample numbers and can be used to ensure that solutions, tubes, tips, etc. are RNase-free; the kit contains sufficient reagents for 25 reactions.
  • Detects as little as 3.5 x 10-7 units (~0.5 pg) of RNase A, with a fluorometer or by eye
  • Simple, straightforward 30 min assay
  • Monitor RNase activity in column fractions during protein purification
  • Use with DNaseAlert™ to quantitatively detect both RNases and DNases in the same sample
Order Info
Shipping Condition: Dry ice
Specifications
Specifications
25 reactions
RNase Control Reagent
For Research Use Only. Not for use in diagnostic procedures.