Please login to your online account to display your discounted pricing

Invitrogen™ Ambion™ RNAqueous Total RNA Isolation Kit DFS Item

Uses glass fiber filter separation technology

$365.00 - $388.00

Specifications

For Use With (Application) Northern Blotting, Nuclease Protection Assays, RT-PCR (Endpoint), Real-Time PCR, cDNA Library Construction
Quantity 50 preps
Sample Type Bacteria, Cells, Plant Samples, Tissue, Viral Samples, Yeast
Target Total RNA
View More Specs
Includes:  6mL Formaldehyde Load Dye, 60mL Lysis/Binding Solution, 60mL Water for 64% Ethanol, 4mL Lithium Chloride Precipitation Solution, 40mL Wash Solution 1, 80mL Wash Solution 2/3 Concentrate, 50 Filter Cartridges, 100 Collection Tubes, 10mL Elution Solution

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Includes Product Type Price Quantity    

AM1912M

DFS Item
 
invitrogen™
LSAM1912M
6mL Formaldehyde Load Dye; 60mL Lysis/Binding Solution; 60mL Water for 64% Ethanol; 4mL Lithium Chloride Precipitation Solution; 40mL Wash Solution 1; 80mL Wash Solution 2/3 Concentrate; 50 Filter Cartridges; 100 Collection Tubes; 10mL Elution Solution; Manual Kit with manual Each for $365.00

AM1912

DFS Item
 
invitrogen™
LSAM1912
6mL Formaldehyde Load Dye; 60mL Lysis/Binding Solution; 60mL Water for 64% Ethanol; 4mL Lithium Chloride Precipitation Solution; 40mL Wash Solution 1; 80mL Wash Solution 2/3 Concentrate; 50 Filter Cartridges; 100 Collection Tubes; 10mL Elution Solution Kit Each for $388.00
Description & Specifications

Specifications

For Use With (Application) Northern Blotting, Nuclease Protection Assays, RT-PCR (Endpoint), Real-Time PCR, cDNA Library Construction
Quantity 50 preps
Sample Type Bacteria, Cells, Plant Samples, Tissue, Viral Samples, Yeast
Target Total RNA

The RNAqueous Kit is for phenol-free total RNA isolation from 0.1g to 0.5g of tissue or from 107 to 108 cells, using a guanidinium-based lysis/denaturant and glass fiber filter separation technology.

  • Convenient syringe-based filtration
  • Compatibility with vacuum manifolds when processing multiple samples
How it works
The RNAqueous Kit uses a powerful chaotropic solution for cell lysis, followed by RNA binding to glass fiber housed in a luer-lock syringe filter. Washing and elution solutions are simply pushed through the filter using a standard syringe (not included). Vacuum filtration is not required; however, the filter units are compatible with standard vacuum manifolds for processing several samples at once.

Not high-throughput compatible (Manual)

Applications:

DNA and RNA Purification and Analysis, RNA Extraction, Total RNA Isolation, Total RNA from Animal Cells and Tissues

Shipping Condition: Room temperature