Please login to your online account to display your discounted pricing

Invitrogen™ Ambion™ RNAqueous Total RNA Isolation Kit DFS Item


Uses glass fiber filter separation technology

Manufacturer: invitrogen™  LSAM1912M

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. AM1912M

  • / Each
 

Description & Specifications

Specifications

Includes 6mL Formaldehyde Load Dye; 60mL Lysis/Binding Solution; 60mL Water for 64% Ethanol; 4mL Lithium Chloride Precipitation Solution; 40mL Wash Solution 1; 80mL Wash Solution 2/3 Concentrate; 50 Filter Cartridges; 100 Collection Tubes; 10mL Elution Solution; Manual
Sample Type Bacteria, Cells, Plant Samples, Tissue, Viral Samples, Yeast
Final Product Type Total RNA
For Use With (Application) Northern Blotting, Nuclease Protection Assays, RT-PCR (Endpoint), Real-Time PCR, cDNA Library Construction
Target Total RNA
Isolation Technology Spin Column (Glass Fiber Filter)
Quantity 50 preps
Product Type Kit with manual
Quantity 50 preps