Please login to your online account to display your discounted pricing

Invitrogen™ Ambion™ RNAqueous-Midi Total RNA Isolation Kit DFS Item


RNA isolation for midrange amounts of starting material

Manufacturer: invitrogen™  LSAM1911M

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. AM1911M

  • / Each
 

Description & Specifications

Specifications

Includes Formaldehyde Load Dye, Lysis/Binding Solution, Water for 64% Ethanol, Lithium, Chloride Solution, Wash Solution No. 1 and Wash Solution 2/3 Concentrate, Syringe Filter Units, Collection Tubes and Elution Solution, Manual
Product Type Kit with manual
Target Total RNA
Isolation Technology Syringe Filter (Glass Fiber)
Quantity 15 preps
For Use With (Application) Northern Blotting, Nuclease Protection Assays, RT-PCR (Endpoint), Real-Time PCR, cDNA Library Construction
Sample Type Tissue Cells
Final Product Type Total RNA