Please login to your online account to display your discounted pricing

Invitrogen™ Ambion™ RNAqueous-Midi Total RNA Isolation Kit DFS Item


RNA isolation for midrange amounts of starting material

Manufacturer: invitrogen™  LSAM1911

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. AM1911

  • / Each
 

Description & Specifications

Specifications

Sample Type Tissue Cells
Final Product Type Total RNA
For Use With (Application) Northern Blotting, Nuclease Protection Assays, RT-PCR (Endpoint), Real-Time PCR, cDNA Library Construction
Target Total RNA
Includes Formaldehyde Load Dye, Lysis/Binding Solution, Water for 64% Ethanol, Lithium, Chloride Solution, Wash Solution No. 1 and Wash Solution 2/3 Concentrate, Syringe Filter Units, Collection Tubes and Elution Solution
Isolation Technology Syringe Filter (Glass Fiber)
Quantity 15 preps
Product Type Kit
Quantity 15 preps