Please login to your online account to display your discounted pricing

Invitrogen™ Ambion™ RNA Rapid Extraction Solution DFS Item

Fast procedure to release RNA from Whatman FTA cards

$175.00 - $201.00

Specifications

For Use With (Application) Northern Blotting, RT-PCR (Endpoint), Real-Time PCR
Quantity 10mL
Storage Requirements Store at room temperature.
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Includes Price Quantity    

AM9775M

DFS Item
 
invitrogen™
LSAM9775M
Solution, with manual Each for $201.00

AM9775

DFS Item
 
invitrogen™
LSAM9775
Solution, without manual Each for $175.00
Description & Specifications

Specifications

For Use With (Application) Northern Blotting, RT-PCR (Endpoint), Real-Time PCR
Quantity 10mL
Storage Requirements Store at room temperature.

Ambion Whatman™ FTA™ Cards are frequently used for the collection, transport, and storage of RNA and DNA in biological samples. The RNA Rapid Extraction Solution is to release RNA from the paper prior to RNA isolation. 10mL is provided. Suitable for use with northern blotting, RT-PCR (endpoint), and real-time PCR.

  • Rapid 5 minutes procedure to release RNA from Whatman FTA cards
  • Ideal for high-throughput applications
  • Integrity of RNA is unaffected

Ready for real-time PCR Disks punched from FTA Cards are simply dropped into RNA Rapid Extraction Solution and agitated for 5 minutes to release the sample for RNA isolation—while protecting the RNA from degradation. The RNA can then be purified from the RNA Rapid Extraction Solution using the MagMAX™-96 Viral RNA Isolation Kit (Cat. No. AM1836). The high-quality purified RNA is suitable for quantitative RT-PCR or other downstream RNA applications. Whatman FTA Cards can be used with virtually any sample type including blood, cultured cells, buccal cells, plant material, bacteria, plasmids, microorganisms, solid tissue, viral particles, and more.

Animal Health, Animal Health Sample Preparation, DNA and RNA Purification and Analysis, RNA Extraction, Total RNA Isolation, Total RNA from Animal Cells and Tissues, Total RNA from Bacterial Samples, Total RNA from Blood, Total RNA from Plant Cells, Viral RNA/DNA Purification

Shipping Condition: Room temperature