Please login to your online account to display your discounted pricing

Invitrogen™ Ambion™ RNA Rapid Extraction Solution DFS Item


Fast procedure to release RNA from Whatman FTA cards

Manufacturer: invitrogen™  LSAM9775M

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. AM9775M

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Northern Blotting, RT-PCR (Endpoint), Real-Time PCR
Quantity 10mL
Includes Solution, with manual
Storage Requirements Store at room temperature.