Please login to your online account to display your discounted pricing

Ambion™ RiboPure™ RNA Purification Kit DFS Item


Enzymatic pretreatment not required

Manufacturer: life technologies  LSAM1924M

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. AM1924M

  • / Each
 

Description & Specifications

Specifications

Final Product Type Total RNA
For Use With (Application) In Vitro Translation, Microarray Analysis, Northern Blotting, Nuclease Protection Assays, RT-PCR (Endpoint), Real-Time PCR, cDNA Library Construction
Yield 500μg
Target Total RNA
Includes TRI Reagent, filter cartridges, 2mL collection tubes, wash solution concentrate and elution buffer, manual
Isolation Technology Organic Extraction, Spin Column
Product Type Kit with manual
Quantity 50 preps
Sample Size Up to 100mg tissue, Up to 2 x 107 cells
Sample Type Cells, Tissue
Quantity 50 preps