Please login to your online account to display your discounted pricing

Invitrogen™ Ambion™ Reservoir DFS Item

For use with Tempus™ blood RNA tubes

$69.00 - $84.79

Specifications

Product Type Reservoir
For Use With (Equipment) Tempus™ blood RNA tubes
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Volume (Metric) Quantity Price Quantity    

4344435

DFS Item
 
life technologies
LS4344435
Each for $69.00

4344437

DFS Item
 
life technologies
LS4344437
Each for $84.79
Description & Specifications

Specifications

Product Type Reservoir
For Use With (Equipment) Tempus™ blood RNA tubes

Reservoirs available in 5mL and 20mL sizes can be used during RNA sample preparation of large volume of blood collected in the Tempus™ blood RNA tubes.

DNA and RNA Purification and Analysis, General RNA Purification Reagents and Accessories, RNA Extraction, Total RNA Isolation, Total RNA from Blood

Shipping Condition: Room temperature