Please login to your online account to display your discounted pricing

Invitrogen™ Ambion™ QuantumRNA™ Universal 18S Internal Standard DFS Item

Used for quantitative end-point RT-PCR

$254.00 - $254.00

Specifications

Quantity 100 reactions
Storage Requirements Store 18S Primer Pair and 18S Competimers at -20°C. Store Control Template RNA below -70°C. Store Nuclease-free water at any temperature.
View More Specs
Includes:  18S Primer Pair and 18S Competimer, Control Template RNA, Nuclease-free water

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Includes Price Quantity    

AM1718

DFS Item
 
invitrogen™
LSAM1718
Kit Each for $254.00
Description & Specifications

Specifications

Quantity 100 reactions
Storage Requirements Store 18S Primer Pair and 18S Competimers at -20°C. Store Control Template RNA below -70°C. Store Nuclease-free water at any temperature.

The Ambion QuantumRNA™ 18S Internal Standard with Competimer™ technology uses specially modified Competimers of the same sequence as the normal 18S primers that cannot be extended. By adjusting the ratio of 18S Competimers to normal 18S mRNA primers, the signal for 18S mRNA can be attenuated to the level of very rare messages. They are used for quantitative end-point RT-PCR and include sufficient reagents for 100 reactions. Because of its invariant expression across tissues and treatments, 18S ribosomal RNA is an ideal internal control for quantitative RNA analysis. The Universal 18S Internal Standards function across the broadest range of eukaryotes, including animals, plants, and many protozoa and produce a 315 bp PCR product.

  • Species: Mouse

Accessory Products:

  • The QuantumRNA Classic 18S Internal Standard (Cat. No. AM1716) produce a different 489 bp PCR product, and the QuantumRNA Classic II 18S Internal Standard (Cat. No. AM1717) produce a 324 bp PCR product.

PCR and Real-Time PCR

Shipping Condition: Dry Ice