Please login to your online account to display your discounted pricing
Invitrogen™ Ambion™ QuantumRNA™ Classic 18S Internal Standard DFS Item
Used for quantitative end-point RT-PCR
Manufacturer: invitrogen™  LSAM1716M
 View more versions of this product
 Disclaimers
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. AM1716M
  • / Each
Description & Specifications
Specifications
Includes Kit with manual
Quantity 100 reactions
Storage Requirements Store 18S Primer Pair and 18S Competimers at –20°C. Store Control Template RNA below –70°C. Store Nuclease-free water at any temperature.