Invitrogen™ Ambion™ QuantumRNA™ β-Actin Internal Standards DFS Item
Used for quantitative end-point RT-PCR
Manufacturer:  Invitrogen™ LSAM1720
 View more versions of this product
Catalog No. AM1720
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specifications Description & Specifications
Specifications
Kit
100 reactions
Store β-Actin Primer Pair and β-Actin Competimers at -20°C. Store Control Template RNA below -70°C. Store Nuclease-free water at any temperature.