Learn More
Invitrogen™ PureLink™ RNA Mini Kit
Provides a simple, reliable, and rapid column-based method for isolating high-quality total RNA from a wide variety of sources without the need for hazardous reagents such as phenol
Manufacturer: Invitrogen™ LS12183018A
Description
Rapid and convenient column purification
The PureLink™ RNA Mini Kit provides rapid purification of total RNA from a wide range of cells and tissue types to yield up to 1000μg of purified RNA from a single extraction. High-quality total RNA can be obtained from mini- to midi-prep amounts of starting material with no genomic DNA contamination.
Simplified, nontoxic RNA purification
The PureLink™ RNA Mini Kit combines nontoxic guanidine-isothiocyanate lysis with the speed, purity, and ease-of-use of silica-membrane purification. The safe and easy procedure can typically be completed in less than 20 minutes without the need for hazardous phenol/chloroform extraction, CsCl centrifugation, LiCl, or alcohol precipitation.
- Obtain high-yield, high-integrity RNA
- Rapid, non-hazardous RNA purification capabilities
- Ready-to-use RNA for many downstream applications
Specifications
Total RNA | |
Total RNA | |
Spin Column | |
50 preps | |
Not High-throughput Compatible (Manual) | |
Up to 200 mg tissue, 5x10^6 to 1 x 10^8 cells | |
PureLink™ |
Microarray Analysis, Next-Generation Sequencing, Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), Nucleic Acid Labeling, Nuclease Protection Assays, Northern Blotting, cDNA Library Construction | |
Liquid Samples (e.g. Serum), Plant Samples, Cells, Tissue, Yeast, Bacteria, Blood | |
Silica Spin Column | |
• Lysis Buffer (125 ml) • Wash Buffer I (50 ml) • Wash Buffer II (15 ml) • RNase-Free Water (15.5 ml) • Spin Cartridges with collection tubes (50 each) • Collection Tubes (50 each) • Recovery Tubes (50 each) Store at room temperature. |
|
Room Temperature | |
PureLink™ |
Promotions
-
Get Gene Expression Products with Purchase
Spend up to $5,000 on select gene expression products and get a matching amount at no additional cost.: 06/30/21
For Research Use Only. Not for use in diagnostic procedures.