missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Ambion™ Phenol:Chloroform:IAA, 25:24:1, pH 6.6 DFS Item

Ready-to-use, safer than preparing crystalline phenol solutions

$105.00 - $215.00

Specifications

Storage Requirements Store at +4° or -20°C.
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AM9730 Invitrogen™
LSAM9730
100mL Each for $105.00
AM9732 Invitrogen™
LSAM9732
400mL Each for $215.00
Specifications

Description

Ambion Acid Phenol:Chloroform:IAA (25:24:1) is premixed and supplied at pH 6.6 (pH 7.9 ±0.2), with included buffer. In RNA and DNA extraction procedures, Acid Phenol:Chloroform:IAA (pH 7.9) helps to stabilize the interface and prevents foaming when mixing. Preparation of phenol for use in molecular biology applications is a time-consuming and often hazardous procedure due to its toxic and corrosive nature. Ambion premixed, quality tested, saturated phenols are ready-to-use, eliminating handling problems and offering a more convenient, safer, and easier alternative to preparing solutions from crystalline phenol.

DNA and RNA Purification and Analysis, DNA Extraction, General RNA Purification Reagents and Accessories, General gDNA Purification Reagents and Accessories, Genomic DNA Purification, RNA Extraction, Total RNA Isolation, Total RNA from Animal Cells and Tissues

Order Info

Shipping Condition: Room temperature

Description & Specifications

Specifications

Store at +4° or -20°C.