Please login to your online account to display your discounted pricing

Invitrogen™ Ambion™ mRNA CATCHER™ PLUS Purification Kit DFS Item


LNA technology offers improved and faster performance

Manufacturer: invitrogen™  LSK157002

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. K157002

  • / Each
 

Description & Specifications

Specifications

Sample Type Blood, Cells, RNA, Tissue
Final Product Type mRNA
For Use With (Equipment) Robotic Liquid Handling Systems
Target mRNA
No. of Samples 96.0
Isolation Technology Oligo dT Affinity
Quantity 96 preps
Test Time 1.5 hrs
Format 96-well plate
For Use With (Application) Microarray Analysis, RT-PCR (Endpoint), Real-Time PCR, cDNA Library Construction
Format 96-well plate
Quantity 96 preps