Please login to your online account to display your discounted pricing

Ambion™ Mouse RiboPure™-Blood RNA Isolation Kit DFS Item


High-quality RNA isolation from mouse and rat blood

Manufacturer: life technologies  LSAM1951M

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. AM1951M

  • / Each
 

Description & Specifications

Specifications

Sample Type Blood
Target Total RNA, miRNA
For Use With (Application) Microarray Analysis, Northern Blotting, RT-PCR (Endpoint), Real-Time PCR
Species Mouse
Product Type Kit with manual
Sample Size Up to 0.5mL whole blood
Yield 4μg
Includes Acid–Phenol:Chloroform, 3M sodium acetate pH 5.5, wash solution 1 concentrate, wash solution 2/3 concentrate, 2mL tubes pre-loaded with RNAlater, lysis solution, filter cartridges, collection tubes, nuclease-free water, manual
Final Product Type Total RNA, micro RNA
Isolation Technology Organic Extraction, Spin Column
Quantity 25 preps