missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Ambion™ mMESSAGE mMACHINE™ T7 ULTRA Transcription Kit DFS Item

Generate RNA transcripts with higher protein yields

$580.00 - $2,270.00

Specifications

For Use With (Application) Gene Expression Analysis and Genotyping, in vitro Transcription
Includes T7 Enzyme Mix, 10X Reaction Buffer, 2XNTP/ARCA Solution, GTP Solution, pTRI-Xef, TURBO DNase, Ammonium Acetate Stop Solution, Lithium Chloride Precipitation Solution, E-PAP, 5X E-PAP Buffer, ATP Solution, MnCl2, Formaldehyde Load Dye, and Nuclease-free Water
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: T7 Enzyme Mix, 10X Reaction Buffer, 2XNTP/ARCA Solution, GTP Solution, pTRI-Xef, TURBO DNase, Ammonium Acetate Stop Solution, Lithium Chloride Precipitation Solution, E-PAP, 5X E-PAP Buffer, ATP Solution, MnCl2, Formaldehyde Load Dye, Nuclease-free Water


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AMB13455 Invitrogen™
LSAMB13455
50 reactions Each for $2,270.00
AM1345 Invitrogen™
LSAM1345
10 reactions Each for $594.00
AM1345M Invitrogen™
LSAM1345M
10 reactions Each for $580.00
Specifications

Description

The mMESSAGE mMACHINE T7 Ultra Kit combines new cap analog, anti-reverse cap analog (ARCA), with patented high-yield transcription technology to generate RNA transcripts that produce higher protein yields compared to other transcripts upon translation.

  • RNA Yield: 20-30μg
  • Synthesize more protein from in vitro-transcribed RNA
  • Express RNA transcripts more efficiently both in vitro and in vivo
  • Stabilize transcripts in vivo with included reagents for poly(A) tailing

Description & Specifications

Specifications

Gene Expression Analysis and Genotyping, in vitro Transcription
T7 Enzyme Mix, 10X Reaction Buffer, 2XNTP/ARCA Solution, GTP Solution, pTRI-Xef, TURBO DNase, Ammonium Acetate Stop Solution, Lithium Chloride Precipitation Solution, E-PAP, 5X E-PAP Buffer, ATP Solution, MnCl2, Formaldehyde Load Dye, and Nuclease-free Water