Please login to your online account to display your discounted pricing

Invitrogen™ Ambion™ mMESSAGE mMACHINE™ T7 Transcription Kit DFS Item


in vitro synthesis of large amounts of capped RNA

Manufacturer: invitrogen™  LSAM1344M

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. AM1344M

  • / Each
 

Description & Specifications

Specifications

Product Type Kit with manual
Concentration 40mM
Includes T7 Enzyme Mix, 10X Reaction Buffer, 2XNTP/CAP Solution, GTP Solution, pTRI-Xef, TURBO DNase, Ammonium Acetate Stop Solution, Lithium Chloride Precipitation Solution, Gel Loading Buffer II, and Nuclease-free Water
Quantity 25 reactions
For Use With (Application) Gene Expression Analysis and Genotyping, in vitro Transcription