Please login to your online account to display your discounted pricing
Invitrogen™ Ambion™ mMESSAGE mMACHINE™ SP6 Transcription Kit DFS Item
in vitro synthesis of large amounts of capped RNA
$584.00 - $592.00
Specifications
For Use With (Application) Gene Expression Analysis and Genotyping, in vitro Transcription
Includes SP6 Enzyme Mix, 10X Reaction Buffer, 2XNTP/CAP Solution, GTP Solution, pTRI-Xef, TURBO DNase, Ammonium Acetate Stop Solution, Lithium Chloride Precipitation Solution, Gel Loading Buffer II, and Nuclease-free Water
Quantity 25 reactions
View More Specs
Includes:  SP6 Enzyme Mix, 10X Reaction Buffer 2XNTP/CAP Solution, GTP Solution, pTRI-Xef, TURBO DNase, Ammonium Acetate Stop Solution, Lithium Chloride Precipitation Solution, Gel Loading Buffer II, Nuclease-free Water
 Disclaimers
For Research Use Only. Not for use in diagnostic procedures.
Products
Catalog Number Mfr. No. Price Quantity    
AM1340
DFS Item
invitrogen™
LSAM1340
Each for $592.00
AM1340M
DFS Item
invitrogen™
LSAM1340M
Each for $584.00
Description & Specifications
Specifications
For Use With (Application) Gene Expression Analysis and Genotyping, in vitro Transcription
Includes SP6 Enzyme Mix, 10X Reaction Buffer, 2XNTP/CAP Solution, GTP Solution, pTRI-Xef, TURBO DNase, Ammonium Acetate Stop Solution, Lithium Chloride Precipitation Solution, Gel Loading Buffer II, and Nuclease-free Water
Quantity 25 reactions
mMESSAGE mMACHINE™ kits are designed for in vitro synthesis of large amounts of capped RNA. Capped RNA mimics most eukaryotic mRNAs found in vivo, because it has a 7-methyl guanosine cap structure at the 5' end.
  • RNA Yield: 5-25μg
  • Include cap analog [m7G(5')ppp(5')G] in an ultra high-yield transcription reaction
  • Cap analog is incorporated only as first or 5' terminal G of transcript because its structure precludes its incorporation at other position in RNA molecule
  • Simplified reaction format in which all four ribonucleotides and cap analog are mixed in single solution
  • Ideal for routine synthesis of capped RNAs for oocyte microinjection, in vitro translation, transfection, and other applications