Please login to your online account to display your discounted pricing
Invitrogen™ Ambion™ Microcentrifuge Spin Columns DFS Item
Sterile, RNase-free
$514.00 - $1,854.00
Specifications
Format Spin Column
View More Specs
Includes:  0.45 micron cellulose acetate microcentrifuge spin columns and sterile microcentrifuge tubes
 Disclaimers
For Research Use Only. Not for use in diagnostic procedures.
Products
Catalog Number Mfr. No. Quantity Price Quantity    
R50050
DFS Item
invitrogen™
LSR50050
50 columns Each for $514.00
R50020
DFS Item
invitrogen™
LSR50020
200 columns Each for $1,854.00
Description & Specifications
Specifications
Format Spin Column
The microcentrifuge spin columns included with the FastTrack™ 2.0 and Micro-FastTrack 2.0 Kits are available separately. The columns are made from polypropylene and are designed to enhance the speed and efficiency of polyA+ RNA isolation. The columns are sterilized and RNase-free.
DNA and RNA Purification and Analysis, RNA Extraction, mRNA Isolation