Please login to your online account to display your discounted pricing
Invitrogen™ Ambion™ MessageAmp™ Premier RNA Amplification Kit DFS Item
Innovative kit for the preparation of RNA samples for microarray analysis
Manufacturer: invitrogen™  LSAM1792
 View more versions of this product
 Disclaimers
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. AM1792
  • / Each
Description & Specifications
Specifications
Quantity 30 reactions
For Use With (Application) Expression Array Labeling, Gene Expression Analysis and Genotyping, Microarray Analysis, RNA Amplification
Includes 33μL First Strand Enzyme Mix, 132μL First Strand Buffer Mix, 66μL Second Strand Enzyme Mix, 165μL Second Strand Buffer Mix, 198μL T7 Enzyme Mix, 660μL T7 Biotin IVT Mix, 10μL Control RNA 1mg/mL, 1.75mL Nuclease-free Water, 1.8mL aRNA Binding Buffer Concentrate, 360μL RNA Binding Beads, 10mL aRNA Wash Solution Concentrate, 5mL aRNA Elution Solution, 1mL 5X Array Fragmentation Buffer, 20 8-Strip PCR Tubes and Caps, 2 U-Bottom Plates, 1 Reservoir, 10mL Nuclease-free Water
Product Type Kit, RNA Amplification