Please login to your online account to display your discounted pricing
Invitrogen™ Ambion™ MessageAmp™ III RNA Amplification Kit DFS Item
Provides extensive improvements over existing methods: fewer components, pipetting steps, and manipulations in the workflow
Manufacturer: invitrogen™  LSAM1793
 View more versions of this product
 Disclaimers
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. AM1793
  • / Each
Description & Specifications
Specifications
Quantity 30 reactions
Product Type Kit, RNA Amplification
Includes The MessageAmp III aRNA Amplification Kit includes First-Strand Enzyme Mix, First-Strand Buffer Mix, Second-Strand Enzyme Mix, Second-Strand Buffer Mix, T7 Enzyme Mix, T7 Biotin IVT Mix, Control RNA (1mg/mL HeLa total RNA), Wash Buffer, aRNA Binding Buffer, Filter Cartridges, Collection Tubes, 8-Strip PCR Tubes and Caps (0.2mL), 5X Array Fragmentation Buffer, and Nuclease-free Water
For Use With (Application) Expression Array Labeling, Gene Expression Analysis and Genotyping, Microarray Analysis, RNA Amplification