Please login to your online account to display your discounted pricing
Invitrogen™ Ambion™ MEGAshortscript™ T7 Transcription Kit DFS Item
Maximize the yield of short RNA transcripts
Manufacturer: invitrogen™  LSAM1354M
 View more versions of this product
 Disclaimers
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. AM1354M
  • / Each
Description & Specifications
Specifications
Storage Requirements T7 Enzyme Mix, 10X Reaction Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, pTRI-RNA 18S, TURBO DNase, Ammonium Acetate Stop Solution, and Gel Loading Buffer II are all stored at -20°C. Nuclease-free Water may be stored at any temperature.
Product Type Kit with manual
For Use With (Application) Gene Expression Analysis and Genotyping, in vitro Transcription
Quantity 25 reactions
Quantity 25 reactions
Includes T7 Enzyme Mix, 10X Reaction Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, pTRI-RNA 18S, TURBO DNase, Ammonium Acetate Stop Solution, Gel Loading Buffer II, and Nuclease-free Water
Product Name Kit with manual