Please login to your online account to display your discounted pricing
Invitrogen™ Ambion™ MEGAscript™ T7 Transcription Kit DFS Item
Ultra-high yield technology for in vitro transcription
Manufacturer: invitrogen™  LSAM1334M
 View more versions of this product
 Disclaimers
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. AM1334M
  • / Each
Description & Specifications
Specifications
For Use With (Application) Gene Expression Analysis and Genotyping, in vitro Transcription
Includes T7 Enzyme Mix, 10X Reaction Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, pTRI-Xef, TURBO DNase, Ammonium Acetate Stop Solution, Lithium Chloride Precipitation Solution, Gel Loading Buffer II, and Nuclease-free Water
Product Type Kit with manual
Quantity 40 reactions
Product Name Kit with manual
Quantity 40 reactions