Invitrogen™ Ambion™ MEGAscript™ T7 Transcription Kit DFS Item
Ultra-high yield technology for in vitro transcription
Manufacturer:  Invitrogen™ LSAM1333M
 View more versions of this product
Catalog No. AM1333M
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specifications Description & Specifications
Specifications
Gene Expression Analysis and Genotyping, in vitro Transcription
T7 Enzyme Mix, 10X Reaction Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, pTRI-Xef, TURBO DNase, Ammonium Acetate Stop Solution, Lithium Chloride Precipitation Solution, Gel Loading Buffer II, and Nuclease-free Water
Kit with manual
25 reactions