missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Ambion™ MEGAscript™ T3 Transcription Kit DFS Item

Synthesize 10 to 50 times the amount of RNA produced by conventional transcription reactions

$397.00 - $415.00

Specifications

For Use With (Application) Gene Expression Analysis and Genotyping, in vitro Transcription
Includes T3 Enzyme Mix, 10X Reaction Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, pTRI-Xef, TURBO DNase, Ammonium Acetate Stop Solution, Lithium Chloride Precipitation Solution, Gel Loading Buffer II, and Nuclease-free Water
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: T3 Enzyme Mix, 10X Reaction Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, pTRI-Xef, TURBO DNase, Ammonium Acetate Stop Solution, Lithium Chloride Precipitation Solution, Gel Loading Buffer II, Nuclease-free Water


Products ${productFamilyLength}
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
AM1338 Invitrogen™
LSAM1338
Each for $415.00
AM1338M Invitrogen™
LSAM1338M
Each for $397.00
Specifications

Description

Novel transcription reaction conditions and Ambion's patented, high-yield technology allow the synthesis of 10 to 50 times the amount of RNA produced by conventional transcription reactions. Each Ambion kit includes sufficient reagents for 40 reactions.

  • Exclusive, ultrahigh yield technology is fast: 2 hour reaction time
  • Amplifies aRNA for gene array analysis and other applications
  • Efficiently incorporates many modified nucleotides
  • A typical 20μL MEGAscript reaction with 1μg of the pTRI-Xef-1α control template will yield over 100μg of transcript
  • Versatility of the MEGAscript kit allows for manipulation of the reactions to include specialized reagents such as modified nucleotides, cap analog, or additional polymerase
Accessory Products:
MessageAmp™ aRNA Amplification Kits are powered by MEGAscript technology and have been developed especially for the amplification of cRNA prior to array analysis. To purify in vitro transcription products from free nucleotides, buffer components, and enzymes, use MEGAclear™ Kit (Mfr. No. AM1908) and the MEGAclear-96 Kit (Mfr. No. AM1909).

Description & Specifications

Specifications

Gene Expression Analysis and Genotyping, in vitro Transcription
T3 Enzyme Mix, 10X Reaction Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, pTRI-Xef, TURBO DNase, Ammonium Acetate Stop Solution, Lithium Chloride Precipitation Solution, Gel Loading Buffer II, and Nuclease-free Water