Please login to your online account to display your discounted pricing
Invitrogen™ Ambion™ MEGAscript™ T3 Transcription Kit DFS Item
Synthesize 10 to 50 times the amount of RNA produced by conventional transcription reactions
Manufacturer: invitrogen™  LSAM1338M
 View more versions of this product
 Disclaimers
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. AM1338M
  • / Each
Description & Specifications
Specifications
Quantity 40 reactions
Includes T3 Enzyme Mix, 10X Reaction Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, pTRI-Xef, TURBO DNase, Ammonium Acetate Stop Solution, Lithium Chloride Precipitation Solution, Gel Loading Buffer II, and Nuclease-free Water
For Use With (Application) Gene Expression Analysis and Genotyping, in vitro Transcription
Product Type Kit with manual
Product Name Kit with manual
Quantity 40 reactions