missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ MEGAclear™ Transcription Clean-Up Kit DFS Item

Quickly cleanup large-scale transcription reactions

$172.00 - $172.00

Specifications

For Use With (Application) Gene Expression Analysis and Genotyping, in vitro Transcription
View More Specs

Includes: Binding Solution, Wash Solution Concentrate, and 5M Ammonium Acetate, Elution Solution, Filter Cartridges, Collection and Elution Tubes

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AM1908 View Documents Invitrogen™
LSAM1908
20 preps (tubes) Each for $172.00
Description

Description

MEGAclear Kit is designed for quick cleanup of any large-scale transcription reaction. MEGAclear Kit efficiently separates RNA from unincorporated NTPs, enzymes, and buffer components.

•Purifies transcription reactions to produce high-quality RNA, free of unincorporated NTPs, enzymes, and buffer components
High recovery from 1ng to 500μg of input
Ideal for purification of aRNA amplification reactions, nonisotopically labeled RNA probes, and capped RNA transcripts for oocyte injections
Quick protocol—purify RNA in less than 30 minutes

  • Using the MEGAclear Kit: Protocol is quick and easy. Transcription reaction is mixed with Binding Solution, passed over an RNA-binding filter, and washed; an elution step releases the purified RNA, which is ready for any application requiring highly pure synthetic or in vitro-transcribed RNA. For RNA recovery of 70% or greater, single-stranded RNA should be >100 nt and double-stranded RNA should be >200 bp.

Specifications

Specifications

Gene Expression Analysis and Genotyping, in vitro Transcription

For Research Use Only. Not for use in diagnostic procedures.