Please login to your online account to display your discounted pricing
Invitrogen™ Ambion™ MAXIscript™ T7 Transcription Kit DFS Item
Fast synthesis of RNA probes for use in ribonuclease protection assays, in situ hybridization, and blot hybridizations
Manufacturer: invitrogen™  LSAM1312M
 View more versions of this product
 Disclaimers
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. AM1312M
  • / Each
Description & Specifications
Specifications
For Use With (Application) DNA and RNA Purification and Analysis, Gene Expression Analysis and Genotyping, in vitro Transcription, Northern Blotting, Nuclease Protection Assays, Nucleic Acid Gel Electrophoresis and Blotting, Southern Blotting
Includes T7 Enzyme Mix, 10X Transcription Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, TURBO DNase, Gel Loading Buffer II, pTRI-b-actin, and Nuclease-free Water
Product Type Kit with manual
Quantity 30 reactions
Product Name Kit with manual
Storage Requirements T7 Enzyme Mix, 10X Transcription Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, TURBO DNase, Gel Loading Buffer II, and pTRI-b-actin are all stored at -20°C. Nuclease-free Water may be stored at any temperature.
Quantity 30 reactions