Please login to your online account to display your discounted pricing

Invitrogen™ Ambion™ M-MLV Reverse Transcriptase DFS Item

Commonly used in the first step of RT-PCR as well as for primer extension experiments

$147.00 - $322.00

Specifications

For Use With (Application) RT-PCR, Primer extension experiments
Sample Type ssRNA, ssDNA, RNA-DNA Hybrid
Storage Requirements Store at -20deg.C
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Includes Quantity Price Quantity    

AM2043

DFS Item
 
life technologies
LSAM2043
M-MLV Reverse Transcriptase (4000 units); 500μL 10X First Strand Synthesis Buffer 4000 units Each for $147.00

AM2044

DFS Item
 
life technologies
LSAM2044
M-MLV Reverse Transcriptase (10,000 units); 500μL 10X First Strand Synthesis Buffer 10,000 units Each for $322.00
Description & Specifications

Specifications

For Use With (Application) RT-PCR, Primer extension experiments
Sample Type ssRNA, ssDNA, RNA-DNA Hybrid
Storage Requirements Store at -20deg.C

Moloney Murine Leukemia Virus Reverse Transcriptase (M-MLV RT) is derived from a cloned region of the pol gene of M-MLV and isolated from an E. coli strain overexpressing this construct.

  • In the presence of a single-stranded RNA or DNA template, primer, and deoxyribonucleotides, M-MLV Reverse Transcriptase (RT) will extend the oligonucleotide primer, producing a complementary strand

  • Optimal reaction temperature: 37°C
  • Final product size: 7kb or less

PCR & Real-Time PCR, Reverse Transcription

Shipping Condition: Dry ice