Please login to your online account to display your discounted pricing

Ambion™ M-MLV Reverse Transcriptase DFS Item


Commonly used in the first step of RT-PCR as well as for primer extension experiments

Manufacturer: life technologies  LSAM2044

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. AM2044

  • / Each
 

Description & Specifications

Specifications

Temperature (English) Operating 98.6deg.F
Temperature (Metric) Operating 37deg.C
For Use With (Application) RT-PCR, Primer extension experiments
Includes M-MLV Reverse Transcriptase (10,000 units); 500μL 10X First Strand Synthesis Buffer
Quantity 10,000 units
Sample Type ssRNA, ssDNA, RNA-DNA Hybrid
Quantity 10,000 units
Storage Requirements Store at -20deg.C