Please login to your online account to display your discounted pricing

Ambion™ LeukoLOCK™ Fractionation & Stabilization Kit DFS Item


RNA from micro-scale samples

Manufacturer: life technologies  LSAM1933M

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. AM1933M

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Microarray Analysis, Northern Blotting, Nuclease Protection Assays, RT-PCR (Endpoint), Real-Time PCR
Sample Type Blood
Target Total RNA
Final Product Type Cells (Leukocytes)
Isolation Technology Magnetic Bead, Vacuum Filter Column
Yield 20μg
Quantity 20 preps
Includes RNAlater solution, rubber septum caps, white slip connectors, transfer spikes, LeukoLOCK filters, elution solution, 1X PBS, manual
Product Type Kit with manual