Invitrogen™ Human Skeletal Muscle Total RNA DFS Item
Highest quality, microRNA-certified
Supplier:  Invitrogen™ AM7982
Catalog No. AM7982
Description
Description
The highest-quality RNA available from Invitrogen, it is DNase-treated, subjected to unsurpassed quality control standards, and certified to contain small RNAs (miRNA, siRNA, and snRNA). One tube containing 100μg is provided at a concentration of 1mg/mL. Intactness of mRNA, genomic DNA contamination, presence/absence of enzymatic inhibitors, and ribosomal RNA contamination in poly(A) preparations are all important aspects used to judge the quality of FirstChoice™ RNAs. The samples are processed using Invitrogen RNA isolation reagents to produce highly pure, intact RNA. A stringent DNase treatment is performed to ensure that the RNA is ready for use in any downstream application, including RT-PCR. Note: Because is impossible to remove every DNA molecule from an RNA sample, each preparation is tested to ensure that any residual DNA contamination is insignificant. The integrity of the RNA is verified by capillary electrophoresis using the Agilent™ 2100 bioanalyzer.
DNA and RNA Purification and Analysis, RNA Extraction
Order Info
Shipping Condition: Dry ice
Certifications
microRNA-Certified
Specifications
Specifications
1 mg/mL
Human
Ambion™
DNase-Treated
Dry Ice
Store at or below -70°C. Avoid multiple freeze–thaw cycles. Aliquots of the product may be stored short-term at -20°C. DO NOT STORE IN FROST-FREE FREEZER.
Total RNA
microRNA-Certified
Normal Tissue
Skeletal Muscle
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.