Invitrogen™ Ambion™ DNAZap™ PCR DNA Degradation Solutions DFS Item
Instantanly degrades high levels of contaminating DNA and RNA from surfaces
$120.00 - $120.00
Specifications
Quantity 250mL
Reagents DNA Decontamination Reagent
View More Specs
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Products ${productFamilyLength}
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
AM9890 Invitrogen™
LSAM9890
Solution Each for $120.00
Specifications
Description
DNAZap™ Solutions are two solutions that are innocuous when used alone, but become potent nucleic acid degrading reagent when mixed. This mixture is able to instantaneously degrade high levels of contaminating DNA and RNA from surfaces.
  • Completely degrades contaminating DNA and RNA at the level of PCR sensitivity
  • Works on contact
  • Degrades nucleic acid, unlike other products on the market that only act as detergents
  • Ideal for cleaning PCR tubes, PCR machine surfaces, pipettors, lab benches, lab equipment, and microfuge tubes
PCR, PCR and Real-Time PCR, Real Time PCR (qPCR)
Order Info
Shipping Condition: Room temperature
Description & Specifications
Specifications
250mL
DNA Decontamination Reagent