Please login to your online account to display your discounted pricing
Invitrogen™ Ambion™ DNA-free™ DNA Removal Kit DFS Item
Safely eliminate DNA contamination from RNA samples
Manufacturer: invitrogen™  LSAM1906M
 View more versions of this product
 Disclaimers
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. AM1906M
  • / Each
Description & Specifications
Specifications
Product Name Kit with manual
Includes RNase-free DNase I, Optimized 10X Reaction Buffer, and a novel DNase Removal Reagent
Quantity 50 reactions
Storage Requirements Store rDNase I, 10X DNase I Buffer and DNase Inactivation Reagent at -20°C. Store Nuclease-free Water at room temperature.
Quantity 50 reactions