Please login to your online account to display your discounted pricing

Invitrogen™ Ambion™ DNA-free™ DNA Removal Kit DFS Item

Safely eliminate DNA contamination from RNA samples

$143.85 - $151.00

Specifications

Storage Requirements Store rDNase I, 10X DNase I Buffer and DNase Inactivation Reagent at -20°C. Store Nuclease-free Water at room temperature.
View More Specs
Includes:  Kit comes complete with RNase-free DNase I, an optimized 10X Reaction Buffer, and a novel DNase Removal Reagent.

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Product Name Price Quantity    

AM1906M

DFS Item
 
invitrogen™
LSAM1906M
Kit with manual Each for $143.85

AM1906

DFS Item
 
invitrogen™
LSAM1906
Kit Each for $151.00
Description & Specifications

Specifications

Storage Requirements Store rDNase I, 10X DNase I Buffer and DNase Inactivation Reagent at -20°C. Store Nuclease-free Water at room temperature.

DNA-free™ DNase treatment and removal reagents are designed for removal of contaminating DNA from RNA samples and for removal of DNase after treatment.

  • No organic extraction or heat inactivation required
  • Includes novel reagent to remove DNase
  • Recombinant DNase I is certified RNase-freeThe DNA-free

  • Inactivation and removal of DNase
  • DNA-free reagents effectively remove DNase and divalent cations from reaction mixture
  • DNase/cation removal step takes only three minutes
  • No organic extraction, EDTA addition, or heat inactivation is required

TURBO™ DNA-free™ Kit is similar to the DNA-free Kit but includes TURBO DNase, an engineered hyperactive DNase that exhibits up to 350% greater catalytic efficiency than wild type DNase. The enzyme also has a 6-fold lower Km for DNA, thus enabling effective removal of trace quantities of DNA contamination.

PCR & Real-Time PCR, Real Time PCR (qPCR), Reverse Transcription

Shipping Conditions: Dry ice